3-Bromothiophene 97%
Supplier: Apollo Scientific
Danger
Synonyms: 3-Thienyl bromide
OR6570-100G OR6570-25G OR6570-500G
APOSOR6570-100GEA 72.5 EUR
APOSOR6570-100G APOSOR6570-25G APOSOR6570-500G
3-Bromothiophene 97%
3-Bromothiophene
Formula: C₄H₃BrS
MW: 163.04 g/mol
Boiling Pt: 92…93 °C/90mmHg
Melting Pt: -10
Density: 1.722 g/cm³ (25 °C)
Flash Pt: 63 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005464
CAS Number: 872-31-1
EINECS: 212-821-3
UN: 2929
ADR: 6.1+3 II