2-Thiopheneethanol
Supplier: Apollo Scientific
Warning
Synonyms: 2-(2-Thienyl)ethanol, Thiophen-2-ethanol
OR6571-100G OR6571-50G OR6571-500G OR6571-25G
APOSOR6571-100GEA 24.8 EUR
APOSOR6571-100G APOSOR6571-50G APOSOR6571-500G APOSOR6571-25G
2-Thiopheneethanol
2-Thiopheneethanol
Formula: C₆H₈OS
MW: 128.19 g/mol
Boiling Pt: 107…109 °C/12mmHg
Density: 1.15 g/cm³ (25 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00005462
CAS Number: 5402-55-1
EINECS: 226-452-0