3-Chlorophenyl acetylene 99%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Chloro-3-ethynylbenzene, 3-Chlorophenylacetylene, (3-Chlorophenyl)acetylene
OR6594-1G OR6594-25G OR6594-5G
APOSOR6594-1GEA 21.9 EUR
APOSOR6594-1G APOSOR6594-25G APOSOR6594-5G
3-Chlorophenyl acetylene 99%
3-Chlorophenyl acetylene
Formula: C₈H₅Cl
MW: 136.578 g/mol
Boiling Pt: 178…179 °C
Density: 1.109 g/cm³ (25 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD03839984
CAS Number: 766-83-6