4-Nitroanisole
Supplier: Apollo Scientific
Warning
OR6596-1KG OR6596-250G
APOSOR6596-1KGEA 132 EUR
APOSOR6596-1KG APOSOR6596-250G
4-Nitroanisole
4-Nitroanisole
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 260 °C
Melting Pt: 50…54 °C
Density: 1.233 g/cm³ (25 °C)
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00007327
CAS Number: 100-17-4
EINECS: 202-825-3
UN: 3458
ADR: 6.1 III
Merck Index: 12,06682