2-Bromo-5-nitroanisole
Supplier: Apollo Scientific
Warning
OR6601-25G OR6601-5G
APOSOR6601-25GEA 33.5 EUR
APOSOR6601-25G APOSOR6601-5G
2-Bromo-5-nitroanisole
2-Bromo-5-nitroanisole
Formula: C₇H₆BrNO₃
MW: 232.03 g/mol
Melting Pt: 102…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00041250
CAS Number: 77337-82-7
EINECS: 278-669-5