Ethyl-4-chloro-2-(methylthio)pyrimidine-5-carboxylate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 4-chloro-2-methylthio-5-pyrimidinecarboxylate, Ethyl 4-chloro-2-(methylsulphanyl)pyrimidine-5-carboxylate, Ethyl 4-chloro-2-(methylthio)pyrimidine-5-carboxylate
OR6605-100G OR6605-25G OR6605-5G
APOSOR6605-100GEA 65.4 EUR
APOSOR6605-100G APOSOR6605-25G APOSOR6605-5G
Ethyl-4-chloro-2-(methylthio)pyrimidine-5-carboxylate 98%
Ethyl 4-chloro-2-(methylthio)pyrimidine-5-carboxylate
Formula: C₈H₉ClN₂O₂S
MW: 232.69 g/mol
Melting Pt: 60…63 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006085
CAS Number: 5909-24-0
EINECS: 227-619-0