2-Nitro-p-toluidine 98+%
Supplier: Apollo Scientific
Danger
OR6639-100G OR6639-2.5KG OR6639-500G
APOSOR6639-100GEA 72.7 EUR
APOSOR6639-100G APOSOR6639-2.5KG APOSOR6639-500G
2-Nitro-p-toluidine 98+%
2-Nitro-p-toluidine
Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Boiling Pt: 169 °C/12mmHg
Melting Pt: 115…118 °C
Density: 1.31 g/cm³ (25 °C)
Flash Pt: 157 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007907
CAS Number: 89-62-3
EINECS: 201-924-9
UN: 2660
ADR: 6.1,III