2-Phenoxyaniline
Supplier: Apollo Scientific
Synonyms: 1-Amino-2-phenoxybenzol
OR6645-25G
APOSOR6645-25GEA 0 EUR
APOSOR6645-25G
2-Phenoxyaniline
2-Phenoxyaniline
Formula: C₁₂H₁₁NO
MW: 185.23 g/mol
Boiling Pt: 170 °C/18mmHg
Melting Pt: 47…49 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00035765
CAS Number: 2688-84-8