N-Phenyl-o-phenylenediamine 98%
Supplier: Apollo Scientific
OR6646-100G OR6646-25G OR6646-10G
APOSOR6646-100GEA 88.6 EUR
APOSOR6646-100G APOSOR6646-25G APOSOR6646-10G
N-Phenyl-o-phenylenediamine 98%
N-Phenyl-o-phenylenediamine
Formula: C₁₂H₁₂N₂
MW: 184.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007685
CAS Number: 534-85-0