4-Amino-6-chloropyrimidine-5-carboxaldehyde
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Amino-6-chloropyrimidine-5-carbaldehyde
OR6659-1G
OR6659-250MG
OR6659-5G
APOSOR6659-1GEA
24.8
EUR
APOSOR6659-1G
APOSOR6659-250MG
APOSOR6659-5G
4-Amino-6-chloropyrimidine-5-carboxaldehyde
4-Amino-6-chloropyrimidine-5-carboxaldehyde
Formula:
C₅H₄ClN₃O Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD03789626 CAS Number: 14160-93-1 EINECS: 238-005-7 |