2,5-Dimethylthiophenol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2,5-Dimethylbenzenethiol, 2,5-Dimethylbenzene-1-thiol, 2-Mercapto-p-xylene, 1,4-Dimethyl-2-mercaptobenzene
OR6661-25G OR6661-5G
APOSOR6661-25GEA 0 EUR
APOSOR6661-25G APOSOR6661-5G
2,5-Dimethylthiophenol 98%
2,5-Dimethylthiophenol
Formula: C₈H₁₀S
MW: 138.23 g/mol
Boiling Pt: 126.3 °C/50mmHg
Density: 1.02 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010020
CAS Number: 4001-61-0
EINECS: 223-649-3
UN: 3334
ADR: 9,III