4-Butoxyaniline 97%

Supplier: Apollo Scientific
Warning

OR6681-25G OR6681-5G
APOSOR6681-25GEA 106 EUR
APOSOR6681-25G APOSOR6681-5G
4-Butoxyaniline 97%
4-Butoxyaniline
Formula: C₁₀H₁₅NO
MW: 165.24 g/mol
Boiling Pt: 131…133 °C/4mmHg
Density: 0.922 g/cm³ (25 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00007866
CAS Number: 4344-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR