(S)-(-)-1,1'-Bi-2-naphthol 99%
Supplier: Apollo Scientific
Danger
Synonyms: (S)-BINOL, (S)-(-)-1,1'-Binaphthyl-2,2'-Diol, (S)-[1,1'-Binaphthalene]-2,2'-diol
OR6693-25G OR6693-5G OR6693-100G
APOSOR6693-25GEA 40.6 EUR
APOSOR6693-25G APOSOR6693-5G APOSOR6693-100G
(S)-(-)-1,1'-Bi-2-naphthol 99%
(S)-(-)-1,1'-Bi-2-naphthol
Formula: C₂₀H₁₄O₂
Melting Pt: 207…210 °C
Storage Temperature: Ambient
MDL Number: MFCD00004068
CAS Number: 18531-99-2
UN: 2811
ADR: 6.1,III