2-(4-Methylsulphonyl-2-nitrophenyl)malondialdehyde 95+%
Supplier: Apollo Scientific
Danger
OR6698-1G OR6698-25G OR6698-5G
APOSOR6698-1GEA 136 EUR
APOSOR6698-1G APOSOR6698-25G APOSOR6698-5G
2-(4-Methylsulphonyl-2-nitrophenyl)malondialdehyde 95+%
2-(4-Methylsulphonyl-2-nitrophenyl)malondialdehyde
Formula: C₁₀H₉NO₆S
MW: 271.25 g/mol
Melting Pt: 213…219 °C
Storage Temperature: Ambient
MDL Number: MFCD00216509
CAS Number: 197251-71-1
UN: 3261
ADR: 8,III