2-Bromo-5-chlorobenzaldehyde
Supplier: Apollo Scientific
|
Warning
|
OR6701-100G
OR6701-25G
OR6701-500G
APOSOR6701-100GEA
94.4
EUR
APOSOR6701-100G
APOSOR6701-25G
APOSOR6701-500G
2-Bromo-5-chlorobenzaldehyde
2-Bromo-5-chlorobenzaldehyde
Formula:
C₇H₄BrClO Melting Pt: 66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00462870 CAS Number: 174265-12-4 |