α,3-Dibromotoluene 97%
Supplier: Apollo Scientific
Danger
Synonyms: 3-Bromobenzyl bromide, 1-Bromo-3-(bromomethyl)benzene
OR6706-100G OR6706-25G OR6706-5G
APOSOR6706-100GEA 0 EUR
APOSOR6706-100G APOSOR6706-25G APOSOR6706-5G
α,3-Dibromotoluene 97%
α,3-Dibromotoluene
Formula: C₇H₆Br₂
MW: 249.93 g/mol
Boiling Pt: 128…130 °C/12mmHg
Melting Pt: 40…42 °C
Density: 1.56 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000176
CAS Number: 823-78-9
EINECS: 212-519-1
UN: 3261
ADR: 8,II