2,6-Dichloro-p-cresol
Supplier: Apollo Scientific
Danger
Synonyms: 2,6-Dichloro-4-methylphenol
OR6709-1G OR6709-25G OR6709-5G
APOSOR6709-1GEA 26.1 EUR
APOSOR6709-1G APOSOR6709-25G APOSOR6709-5G
2,6-Dichloro-p-cresol
2,6-Dichloro-p-cresol
Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
Boiling Pt: 231 °C
Melting Pt: 33…39 °C
Storage Temperature: Ambient
MDL Number: MFCD00134679
CAS Number: 2432-12-4
EINECS: 219-402-4