4-Iodobenzoyl chloride 97%
Supplier: Apollo Scientific
Danger
Synonyms: p-Iodobenzoyl chloride
OR6727-100G OR6727-250G OR6727-25G
APOSOR6727-100GEA 109 EUR
APOSOR6727-100G APOSOR6727-250G APOSOR6727-25G
4-Iodobenzoyl chloride 97%
4-Iodobenzoyl chloride
Formula: C₇H₄ClIO
MW: 266.47 g/mol
Boiling Pt: 120…121 °C/1mmHg
Melting Pt: 61…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00001058
CAS Number: 1711-02-0
EINECS: 216-974-7
UN: 3261
ADR: 8 II