3-Bromosalicylaldehyde 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Bromo-2-hydroxybenzaldehyde
OR6778-25G OR6778-5G OR6778-100G
APOSOR6778-25GEA 119 EUR
APOSOR6778-25G APOSOR6778-5G APOSOR6778-100G
3-Bromosalicylaldehyde 98%
3-Bromosalicylaldehyde
Formula: C₇H₅BrO₂
MW: 201.02 g/mol
Melting Pt: 53…57 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00016587
CAS Number: 1829-34-1