(4-Iodophenyl)acetonitrile 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(4-Iodophenyl)acetonitrile, 4-Iodobenzyl cyanide, 4-Iodophenylacetonitrile
OR6785-1G OR6785-25G OR6785-5G OR6785-100G
APOSOR6785-1GEA 17.6 EUR
APOSOR6785-1G APOSOR6785-25G APOSOR6785-5G APOSOR6785-100G
(4-Iodophenyl)acetonitrile 98%
(4-Iodophenyl)acetonitrile
Formula: C₈H₆IN
MW: 243.05 g/mol
Boiling Pt: 300
Melting Pt: 53…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00060306
CAS Number: 51628-12-7