(3-Methoxyphenyl)acetic acid 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Methoxyphenylacetic acid
OR6816-25G OR6816-5G
APOSOR6816-25GEA 29 EUR
APOSOR6816-25G APOSOR6816-5G
(3-Methoxyphenyl)acetic acid 98%
(3-Methoxyphenyl)acetic acid
Formula: (C₆H₄(OCH₃))CH₂COOH
MW: 166.18 g/mol
Boiling Pt: 305.95
Melting Pt: 70…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00004334
CAS Number: 1798-09-0