Diethyl (cyanomethyl)phosphonate 98%
Supplier: Apollo Scientific
Danger
OR6821-100G OR6821-25G OR6821-250G
APOSOR6821-100GEA 58 EUR
APOSOR6821-100G APOSOR6821-25G APOSOR6821-250G
Diethyl (cyanomethyl)phosphonate 98%
Diethyl (cyanomethyl)phosphonate
Formula: C₆H₁₂NO₃P
MW: 177.14 g/mol
Boiling Pt: 101…102 °C/0.4mmHg
Density: 1.095 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00001893
CAS Number: 2537-48-6
EINECS: 219-806-0
UN: 1760
ADR: 8,II