Cyclopentyltriphenylphosphonium bromide 97%
Supplier: Apollo Scientific
Warning
OR6823-10G OR6823-50G OR6823-250G
APOSOR6823-10GEA 72.5 EUR
APOSOR6823-10G APOSOR6823-50G APOSOR6823-250G
Cyclopentyltriphenylphosphonium bromide 97%
Cyclopentyltriphenylphosphonium bromide
Formula: C₂₃H₂₄BrP
MW: 411.32 g/mol
Melting Pt: 260…262 °C
Storage Temperature: Ambient
MDL Number: MFCD00051880
CAS Number: 7333-52-0