6-Amino-1,4-benzodioxane
Supplier: Apollo Scientific
Warning
Synonyms: 2,3-Dihydrobenzo[b][1,4]dioxin-6-amine, 1,4-Benzodioxan-6-amine
OR6857-100G OR6857-25G OR6857-5G
APOSOR6857-100GEA 372 EUR
APOSOR6857-100G APOSOR6857-25G APOSOR6857-5G
6-Amino-1,4-benzodioxane
6-Amino-1,4-benzodioxane
Formula: C₈H₉NO₂
Boiling Pt: 116…118 °C/3mmHg
Melting Pt: 26…31 °C
Density: 1.231 g/cm³ (25 °C)
Flash Pt: 120 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006824
CAS Number: 22013-33-8
EINECS: 244-718-4