1-(2-Tetrahydrofuroyl)piperazine 97%
Supplier: Apollo Scientific
Warning
Synonyms: (Piperazin-1-yl)(tetrahydrofuran-2-yl)methanone
OR6858-100G OR6858-10G OR6858-25G OR6858-5G
APOSOR6858-100GEA 422 EUR
APOSOR6858-100G APOSOR6858-10G APOSOR6858-25G APOSOR6858-5G
1-(2-Tetrahydrofuroyl)piperazine 97%
1-(2-Tetrahydrofuroyl)piperazine
Formula: C₉H₁₆N₂O₂
Boiling Pt: 120…125 °C/0.2mmHg
Density: 1.17 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00190247
CAS Number: 63074-07-7