2-(1-Piperazinylcarbonyl)-1,4-benzodioxane hydrochloride 98%
Supplier: Apollo Scientific
Danger
Synonyms: 1-[(1,4-Benzodioxan-2-yl)carbonyl]piperazine Hydrochloride, 2-(1-Piperazinylcarbonyl)-1,4-benzodioxane Hydrochloride
OR6859-1G OR6859-250MG
APOSOR6859-1GEA 21.9 EUR
APOSOR6859-1G APOSOR6859-250MG
2-(1-Piperazinylcarbonyl)-1,4-benzodioxane hydrochloride 98%
2-(1-Piperazinylcarbonyl)-1,4-benzodioxane hydrochloride
Formula: C₁₃H₁₆N₂O₃
MW: 284.74 g/mol
Melting Pt: 263 °C
Storage Temperature: Ambient
MDL Number: MFCD00729051
CAS Number: 70918-74-0
UN: 2811
ADR: 6.1,III