1-Benzhydrylpiperazine 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(Diphenylmethyl)piperazine
OR6872-100G OR6872-50G
APOSOR6872-100GEA 24.8 EUR
APOSOR6872-100G APOSOR6872-50G
1-Benzhydrylpiperazine 97%
1-Benzhydrylpiperazine
Formula: C₁₇H₂₀N₂
MW: 252.36 g/mol
Boiling Pt: 185 °C/2.25mmHg
Melting Pt: 88…93 °C
Flash Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00038379
CAS Number: 841-77-0
EINECS: 212-667-7