1-(2-Furoyl)piperazine 96%
Supplier: Apollo Scientific
Warning
Synonyms: Prazosin Related Compound D
OR6882-10G OR6882-25G
APOSOR6882-10GEA 87.3 EUR
APOSOR6882-10G APOSOR6882-25G
1-(2-Furoyl)piperazine 96%
1-(2-Furoyl)piperazine
Formula: C₉H₁₂N₂O₂
MW: 180.21 g/mol
Melting Pt: 67…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00038831
CAS Number: 40172-95-0