1-Nonylpiperazine 98%
Supplier: Apollo Scientific
|
Warning
|
OR6896-10G
OR6896-2G
OR6896-5G
APOSOR6896-10GEA
288
EUR
APOSOR6896-10G
APOSOR6896-2G
APOSOR6896-5G
1-Nonylpiperazine 98%
1-Nonylpiperazine
Formula:
C₁₃H₂₈N₂ Boiling Pt: 105…107 °C/5mmHg Storage Temperature: Ambient |
MDL Number:
MFCD01320897 CAS Number: 82394-25-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...