1-(2-Ethoxyethyl)piperazine 98%
Supplier: Apollo Scientific
Warning
OR6899-1G OR6899-250MG OR6899-5G
APOSOR6899-1GEA 116 EUR
APOSOR6899-1G APOSOR6899-250MG APOSOR6899-5G
1-(2-Ethoxyethyl)piperazine 98%
1-(2-Ethoxyethyl)piperazine
Formula: C₈H₁₈N₂O
Boiling Pt: 224…225 °C
Density: 0.941 g/cm³ (25 °C)
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD01320899
CAS Number: 13484-38-3
UN: 1993
ADR: 3,III