1-(3-Dimethylaminopropyl)piperazine 97%
Supplier: Apollo Scientific
Danger
Synonyms: N,N-Dimethyl-3-(piperazin-1-yl)propan-1-amine
OR6909-1G OR6909-5G
APOSOR6909-1GEA 17.6 EUR
APOSOR6909-1G APOSOR6909-5G
1-(3-Dimethylaminopropyl)piperazine 97%
1-(3-Dimethylaminopropyl)piperazine
Formula: C₉H₂₁N₃
MW: 171.29 g/mol
Boiling Pt: 88…90 °C
Density: 0.917 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00082601
CAS Number: 877-96-3
UN: 2735
ADR: 8 III