1-(2-Piperidin-1-ylethyl)piperazine 98+%
Supplier: Apollo Scientific
Warning
OR6913-1G OR6913-5G OR6913-25G
APOSOR6913-1GEA 109 EUR
APOSOR6913-1G APOSOR6913-5G APOSOR6913-25G
1-(2-Piperidin-1-ylethyl)piperazine 98+%
1-(2-Piperidin-1-ylethyl)piperazine
Formula: C₁₁H₂₃N₃
MW: 197.33 g/mol
Boiling Pt: 119…120 °C/3mmHg
Melting Pt: 60…62 °C
Storage Temperature: Ambient
MDL Number: MFCD01075189
CAS Number: 22763-65-1