4-Hydroxy-1-methylpiperidine 98%
Supplier: Apollo Scientific
Danger
Synonyms: N-Methyl-4-piperidinol, 1-Methyl-4-piperidinol, 1-Methylpiperidin-4-ol
OR6932-25G OR6932-5G
APOSOR6932-25GEA 24.8 EUR
APOSOR6932-25G APOSOR6932-5G
4-Hydroxy-1-methylpiperidine 98%
4-Hydroxy-1-methylpiperidine
Formula: C₆H₁₃NO
MW: 115.18 g/mol
Boiling Pt: 198…200 °C
Melting Pt: 26…31 °C
Density: 0.98 g/cm³ (25 °C)
Flash Pt: 112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006500
CAS Number: 106-52-5
EINECS: 203-406-8