2,2,6,6-Tetramethylpiperidin-4-ol 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2,2,6,6-Tetramethyl-4-piperidinol, 4-Hydroxy-2,2,6,6-tetramethylpiperidine
OR6938-100G OR6938-1KG OR6938-50G
APOSOR6938-100GEA 24.8 EUR
APOSOR6938-100G APOSOR6938-1KG APOSOR6938-50G
2,2,6,6-Tetramethylpiperidin-4-ol 98%
2,2,6,6-Tetramethylpiperidin-4-ol
Formula: C₉H₁₉NO
MW: 157.26 g/mol
Boiling Pt: 212…215 °C
Melting Pt: 129…133 °C
Density: 1.085 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005983
CAS Number: 2403-88-5
EINECS: 219-291-2
UN: 3259
ADR: 8,III