1-Benzyl-4-piperidone 98%
Supplier: Apollo Scientific
Warning
Synonyms: N-Benzyl-4-piperidone, 1-Benzylpiperidin-4-one, 1-Benzyl-4-piperidinone
OR6943-100G OR6943-25G OR6943-500G
APOSOR6943-100GEA 39.3 EUR
APOSOR6943-100G APOSOR6943-25G APOSOR6943-500G
1-Benzyl-4-piperidone 98%
1-Benzyl-4-piperidone
Formula: C₁₂H₁₅NO
Boiling Pt: 248 °C
Density: 1.06 g/cm³ (25 °C)
Flash Pt: 71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006192
CAS Number: 3612-20-2
EINECS: 222-782-4