5-Bromo-2-methylpyridine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Bromo-2-picoline, 3-Bromo-6-picoline
OR6948-100G OR6948-25G OR6948-5G
APOSOR6948-100GEA 77 EUR
APOSOR6948-100G APOSOR6948-25G APOSOR6948-5G
5-Bromo-2-methylpyridine 98%
5-Bromo-2-methylpyridine
Formula: C₆H₆BrN
Melting Pt: 32…36 °C
Storage Temperature: Ambient
MDL Number: MFCD00661170
CAS Number: 3430-13-5