3-Methylsalicylaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 2-Hydroxy-3-methylbenzaldehyde
OR6976-25G OR6976-100G
APOSOR6976-25GEA 92.8 EUR
APOSOR6976-25G APOSOR6976-100G
3-Methylsalicylaldehyde
3-Methylsalicylaldehyde
Formula: C₈H₈O₂
MW: 136.1479 g/mol
Boiling Pt: 209 °C
Melting Pt: 17 °C
Density: 1.123 g/cm³ (25 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00040798
CAS Number: 824-42-0