4-Bromosalicylaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromo-2-hydroxybenzaldehyde
OR6981-5G OR6981-1G OR6981-25G
APOSOR6981-5GEA 37.7 EUR
APOSOR6981-5G APOSOR6981-1G APOSOR6981-25G
4-Bromosalicylaldehyde
4-Bromosalicylaldehyde
Formula: C₇H₅BrO₂
MW: 201.02 g/mol
Melting Pt: 50…54 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD06252606
CAS Number: 22532-62-3
UN: 3077
ADR: 9+EHS III