2-(2-Hydroxycarbonyl-6-pyridyl)malondialdehyde 95+%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(2-Carboxypyridin-6-yl)malondialdehyde
OR6983-1G OR6983-5G OR6983-25G
APOSOR6983-1GEA 136 EUR
APOSOR6983-1G APOSOR6983-5G APOSOR6983-25G
2-(2-Hydroxycarbonyl-6-pyridyl)malondialdehyde 95+%
2-(2-Hydroxycarbonyl-6-pyridyl)malondialdehyde
Formula: C₉H₇NO₄
MW: 193.16 g/mol
Melting Pt: 280 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00216531
CAS Number: 212755-80-1