3-Formylphenylboronic acid
Supplier: Apollo Scientific
Danger
Synonyms: Dihydroxy(3-formylphenyl)borane, 3-Boronobenzaldehyde, (3-Formylphenyl)metaboric acid, 3-Formylbenzeneboronic acid, (3-Formylphenyl)boronic acid
OR6998-100G OR6998-25G
APOSOR6998-100GEA 87 EUR
APOSOR6998-100G APOSOR6998-25G
3-Formylphenylboronic acid
3-Formylphenylboronic acid
Formula: C₇H₇BO₃
MW: 149.94 g/mol
Melting Pt: 109…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00161356
CAS Number: 87199-16-4
UN: 3261
ADR: 8,III