2H-1,4-Benzothiazin-3(4H)-one 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2H-Benzo[b][1,4]thiazin-3(4H)-one
OR7000-100G OR7000-25G
APOSOR7000-100GEA 328 EUR
APOSOR7000-100G APOSOR7000-25G
2H-1,4-Benzothiazin-3(4H)-one 98%
2H-1,4-Benzothiazin-3(4H)-one
Formula: C₈H₇NOS
MW: 165.21 g/mol
Melting Pt: 176…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00006698
CAS Number: 5325-20-2
EINECS: 226-197-5