Methyl-2-chloropyrimidine-5-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 2-chloropyrimidine-5-carboxylate
OR70004-10G OR70004-1G OR70004-5G
APOSOR70004-10GEA 125 EUR
APOSOR70004-10G APOSOR70004-1G APOSOR70004-5G
Methyl-2-chloropyrimidine-5-carboxylate
Methyl-2-chloropyrimidine-5-carboxylate
Formula: C₆H₅ClN₂O₂
MW: 172.5691 g/mol
Melting Pt: 76…80 °C
Storage Temperature: Ambient
MDL Number: MFCD08690330
CAS Number: 287714-35-6
UN: 2811
ADR: 6.1 III