4-Cyano-1-butyne 97%
Supplier: Apollo Scientific
Danger
OR70029-1G OR70029-5G OR70029-25G
APOSOR70029-1GEA 36.4 EUR
APOSOR70029-1G APOSOR70029-5G APOSOR70029-25G
4-Cyano-1-butyne 97%
4-Cyano-1-butyne
Formula: C₅H₅N
MW: 79.999 g/mol
Boiling Pt: 79 °C/35mmHg
Density: 0.88 g/cm³ (25 °C)
Flash Pt: 65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041563
CAS Number: 19596-07-7
UN: 3276
ADR: 6.1 III