tert-Butanol 99%
Supplier: Apollo Scientific
Danger
OR70032-1L OR70032-2.5L
APOSOR70032-1LEA 101 EUR
APOSOR70032-1L APOSOR70032-2.5L
tert-Butanol 99%
tert-Butanol
Formula: (CH₃)₃COH
Boiling Pt: 83 °C
Melting Pt: 23…26 °C
Density: 0.78 g/cm³ (25 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004464
CAS Number: 75-65-0
EINECS: 200-889-7
UN: 1120
ADR: 3,II