4-Bromo-4'-methylbiphenyl
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromo-4'-methyl-1,1'-biphenyl
OR70052-1G OR70052-250MG OR70052-5G
APOSOR70052-1GEA 29 EUR
APOSOR70052-1G APOSOR70052-250MG APOSOR70052-5G
4-Bromo-4'-methylbiphenyl
4-Bromo-4'-methylbiphenyl
Formula: C₁₃H₁₁Br
MW: 247.14 g/mol
Melting Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD00466236
CAS Number: 50670-49-0