2-Bromo-3-nitrophenol 97+%
Supplier: Apollo Scientific
Warning
OR70053-1G OR70053-25G OR70053-5G
APOSOR70053-1GEA 45.1 EUR
APOSOR70053-1G APOSOR70053-25G APOSOR70053-5G
2-Bromo-3-nitrophenol 97+%
2-Bromo-3-nitrophenol
Formula: C₆H₄BrNO₃
MW: 218.0100 g/mol
Melting Pt: 114…116 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08704555
CAS Number: 101935-40-4