Chloro(1,5-cyclooctadiene)rhodium(I) dimer 98%
Supplier: Apollo Scientific
Warning
Synonyms: (1,5-Cyclooctadiene)rhodium chloride dimer
OR70089-1G OR70089-250MG OR70089-100MG
APOSOR70089-1GEA 582 EUR
APOSOR70089-1G APOSOR70089-250MG APOSOR70089-100MG
Chloro(1,5-cyclooctadiene)rhodium(I) dimer 98%
Chloro(1,5-cyclooctadiene)rhodium (I) dimer
Formula: C₁₆H₂₄Cl₂Rh₂
Melting Pt: 243 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012415
CAS Number: 12092-47-6
EINECS: 235-157-6