Tetrabutylammonium hydroxide 1 M methanol
Supplier: Apollo Scientific
Danger
OR70095-100ML OR70095-25ML OR70095-500ML
APOSOR70095-100MLEA 109 EUR
APOSOR70095-100ML APOSOR70095-25ML APOSOR70095-500ML
Tetrabutylammonium hydroxide 1 M methanol
Tetrabutylammonium hydroxide
Formula: C₁₆H₃₇NO
MW: 259.48 g/mol
Melting Pt: 29.85
Density: 0.83 g/cm³ (25 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00009425
CAS Number: 2052-49-5
EINECS: 218-147-6
UN: 2924
ADR: 3+8 II