1-Methyl-5-nitro-1H-indole
Supplier: Apollo Scientific
Warning
OR70135-1G OR70135-5G
APOSOR70135-1GEA 21.9 EUR
APOSOR70135-1G APOSOR70135-5G
1-Methyl-5-nitro-1H-indole
1-Methyl-5-nitro-1H-indole
Formula: C₉H₈N₂O₂
MW: 176.1700 g/mol
Melting Pt: 162…165 °C
Storage Temperature: Ambient
MDL Number: MFCD07772841
CAS Number: 29906-67-0