3-Chlorocatechol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Chloro-2,3-dihydroxybenzene, 3-Chlorobenzene-1,2-diol
OR70152-1G OR70152-250MG OR70152-5G
APOSOR70152-1GEA 122 EUR
APOSOR70152-1G APOSOR70152-250MG APOSOR70152-5G
3-Chlorocatechol 98%
3-Chlorocatechol
Formula: C₆H₅ClO₂
MW: 144.55 g/mol
Boiling Pt: 138 °C/20mmHg
Melting Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00143111
CAS Number: 4018-65-9